Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTAATTTGCATGTGTTAATGC[A/G]TTGATCTATGAGATGGGTATTATGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603820 MIM: 603821 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FFAR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FFAR1 - free fatty acid receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FFAR3 - free fatty acid receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005304.3 | 54 | Intron | NP_005295.1 | |||
XM_011526858.2 | 54 | UTR 5 | XP_011525160.1 |