Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAACCCAGTTACATTTTTGGAGA[C/T]TCTGAAACACTGGAATAGAACTAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604541 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT37 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KRT37 - keratin 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003770.4 | 1666 | UTR 3 | NP_003761.3 |
LOC100505782 - uncharacterized LOC100505782 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |