Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTCCCAGGGATGAGAGCCGAGC[A/C]TGGCTGGGAGGGGATGCAGGCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603150 MIM: 602216 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP5D - ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CBARP - CACN beta subunit associated regulatory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152769.2 | Intron | NP_689982.3 | ||||
XM_017026555.1 | Intron | XP_016882044.1 | ||||
XM_017026556.1 | Intron | XP_016882045.1 | ||||
XM_017026557.1 | Intron | XP_016882046.1 | ||||
XM_017026558.1 | Intron | XP_016882047.1 | ||||
XM_017026559.1 | Intron | XP_016882048.1 | ||||
XM_017026560.1 | Intron | XP_016882049.1 | ||||
XM_017026561.1 | Intron | XP_016882050.1 | ||||
XM_017026562.1 | Intron | XP_016882051.1 |
STK11 - serine/threonine kinase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |