Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGCGGCCGGGGCTGGTCGTGGT[C/G]CATGCAGAGGACGGCACCACCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603150 MIM: 606700 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5D PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP5D - ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001975.1 | 362 | Silent Mutation | GTC,GTG | V,V 87 | NP_001001975.1 | |
NM_001687.4 | 362 | Silent Mutation | GTC,GTG | V,V 87 | NP_001678.1 |
CBARP - CACN beta subunit associated regulatory protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIDN - midnolin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |