Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACAGAGGAACTGAGTAGCTGAA[G/T]GAATGATCCTTTGCTAATTGGTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600940 MIM: 609735 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LIG3 - DNA ligase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002311.4 | 3338 | Intron | NP_002302.2 | |||
NM_013975.3 | 3338 | Intron | NP_039269.2 | |||
XM_005257970.3 | 3338 | Intron | XP_005258027.1 | |||
XM_006721896.3 | 3338 | Intron | XP_006721959.1 | |||
XM_017024624.1 | 3338 | Intron | XP_016880113.1 |
RAD51L3-RFFL - RAD51L3-RFFL readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFFL - ring finger and FYVE-like domain containing E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017368.1 | 3338 | UTR 3 | NP_001017368.1 |
Set Membership: |
HapMap Validated |