Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATTCAAGATTGTACCTTTGGAA[A/G]TGACTCCATTGTGTACCATTCTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601025 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP2B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP2B1 - adaptor related protein complex 2 beta 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030006.1 | Intron | NP_001025177.1 | ||||
NM_001282.2 | Intron | NP_001273.1 | ||||
XM_005257937.3 | Intron | XP_005257994.1 | ||||
XM_005257938.2 | Intron | XP_005257995.1 | ||||
XM_005257941.2 | Intron | XP_005257998.1 | ||||
XM_011524448.1 | Intron | XP_011522750.1 | ||||
XM_011524449.2 | Intron | XP_011522751.1 | ||||
XM_011524450.1 | Intron | XP_011522752.1 | ||||
XM_011524451.1 | Intron | XP_011522753.1 | ||||
XM_011524452.1 | Intron | XP_011522754.1 | ||||
XM_011524453.1 | Intron | XP_011522755.1 | ||||
XM_011524454.1 | Intron | XP_011522756.1 | ||||
XM_011524455.2 | Intron | XP_011522757.1 | ||||
XM_017024284.1 | Intron | XP_016879773.1 | ||||
XM_017024285.1 | Intron | XP_016879774.1 | ||||
XM_017024286.1 | Intron | XP_016879775.1 | ||||
XM_017024287.1 | Intron | XP_016879776.1 |