Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGGCGGTGTTCCTGGTGGACAC[A/G]GGGGATGCCATGAGCCCTGAGCTGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601237 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF112 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RNF112 - ring finger protein 112 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007148.4 | 716 | Silent Mutation | ACA,ACG | T,T 248 | NP_009079.2 | |
XM_006721571.3 | 716 | Silent Mutation | ACA,ACG | T,T 248 | XP_006721634.1 | |
XM_006721572.3 | 716 | Silent Mutation | ACA,ACG | T,T 247 | XP_006721635.1 | |
XM_006721573.3 | 716 | Silent Mutation | ACA,ACG | T,T 224 | XP_006721636.1 |
Set Membership: |
HapMap |