Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605977 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IMMP2L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IMMP2L - inner mitochondrial membrane peptidase subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244606.1 | Intron | NP_001231535.1 | ||||
NM_032549.3 | Intron | NP_115938.1 | ||||
XM_005250630.3 | Intron | XP_005250687.1 | ||||
XM_011516605.2 | Intron | XP_011514907.1 | ||||
XM_011516608.2 | Intron | XP_011514910.1 | ||||
XM_011516609.2 | Intron | XP_011514911.1 | ||||
XM_011516611.2 | Intron | XP_011514913.1 | ||||
XM_011516613.2 | Intron | XP_011514915.1 | ||||
XM_017012699.1 | Intron | XP_016868188.1 | ||||
XM_017012700.1 | Intron | XP_016868189.1 | ||||
XM_017012701.1 | Intron | XP_016868190.1 | ||||
XM_017012702.1 | Intron | XP_016868191.1 | ||||
XM_017012703.1 | Intron | XP_016868192.1 | ||||
XM_017012704.1 | Intron | XP_016868193.1 | ||||
XM_017012705.1 | Intron | XP_016868194.1 |
LRRN3 - leucine rich repeat neuronal 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099658.1 | Intron | NP_001093128.1 | ||||
NM_001099660.1 | Intron | NP_001093130.1 | ||||
NM_018334.4 | Intron | NP_060804.3 |
Set Membership: |
HapMap |