Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGGCCGCCCTCCCTCCTCCCCC[A/G]TCACATTTGAAGTGGTGCATTGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612159 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPH3A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RPH3A - rabphilin 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143854.1 | Intron | NP_001137326.1 | ||||
NM_014954.3 | Intron | NP_055769.2 | ||||
XM_011538048.1 | Intron | XP_011536350.1 | ||||
XM_017019034.1 | Intron | XP_016874523.1 |