Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAAGGGGTCACAGTAGGTTCAT[A/G]GGTGTCATTCTGATTAGAAGTCCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607536 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRTC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CRTC1 - CREB regulated transcription coactivator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098482.1 | Intron | NP_001091952.1 | ||||
NM_015321.2 | Intron | NP_056136.2 | ||||
XM_005259833.3 | Intron | XP_005259890.1 | ||||
XM_005259834.1 | Intron | XP_005259891.1 | ||||
XM_005259835.3 | Intron | XP_005259892.1 | ||||
XM_005259836.3 | Intron | XP_005259893.1 | ||||
XM_006722710.3 | Intron | XP_006722773.1 | ||||
XM_011527842.2 | Intron | XP_011526144.1 | ||||
XM_017026536.1 | Intron | XP_016882025.1 | ||||
XM_017026537.1 | Intron | XP_016882026.1 | ||||
XM_017026538.1 | Intron | XP_016882027.1 |
Set Membership: |
HapMap |