Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCTGAGCCTGTATTTAGAGAG[C/G]GGGGAGGAAAGAGGTCATTGAGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612482 MIM: 603555 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF43 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF43 - ring finger protein 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305544.1 | Intron | NP_001292473.1 | ||||
NM_001305545.1 | Intron | NP_001292474.1 | ||||
NM_017763.5 | Intron | NP_060233.3 | ||||
XM_011524955.2 | Intron | XP_011523257.1 | ||||
XM_011524956.2 | Intron | XP_011523258.1 | ||||
XM_017024800.1 | Intron | XP_016880289.1 |
SUPT4H1 - SPT4 homolog, DSIF elongation factor subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPOAP1-AS1 - TSPOAP1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |