Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAAGGGCCCCAGCTCACAAGTTT[A/T]CGATAGCTTCCTGTGTGGCAGGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604135 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFE2L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NFE2L3 - nuclear factor, erythroid 2 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004289.6 | Intron | NP_004280.5 |
Set Membership: |
Validated |