Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGGAGCAGTCTGAGCTGCTCCT[C/T]CGTCATCCAAGGGGCTCCTCCTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615747 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEACAM8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEACAM8 - carcinoembryonic antigen related cell adhesion molecule 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001816.3 | 862 | Intron | NP_001807.2 | |||
XM_011526340.2 | 862 | Missense Mutation | GAA,GGA | E,G 345 | XP_011524642.1 | |
XM_011526341.1 | 862 | Intron | XP_011524643.1 | |||
XM_011526342.1 | 862 | UTR 3 | XP_011524644.1 | |||
XM_017026194.1 | 862 | Missense Mutation | GAA,GGA | E,G 345 | XP_016881683.1 | |
XM_017026195.1 | 862 | Intron | XP_016881684.1 | |||
XM_017026196.1 | 862 | Intron | XP_016881685.1 | |||
XM_017026197.1 | 862 | Missense Mutation | GAA,GGA | E,G 225 | XP_016881686.1 | |
XM_017026198.1 | 862 | Intron | XP_016881687.1 |
LIPE-AS1 - LIPE antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |