Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTGTGCCAGAGTGGCTCTGTGC[C/T]GGGGTGGCTCTGTGCCGCCGCCGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600038 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MATK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MATK - megakaryocyte-associated tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFR2 - zinc finger RNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145640.1 | Intron | NP_001139112.1 | ||||
NM_015174.1 | Intron | NP_055989.1 | ||||
XM_011527831.2 | Intron | XP_011526133.1 |
Set Membership: |
HapMap Validated |