Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGACCGGAATTCCTTCCAATGA[C/T]GTTTATAAAATCAGGTCTATTTTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUM1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MUM1 - melanoma associated antigen (mutated) 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032853.3 | Intron | NP_116242.2 | ||||
XM_011528380.2 | Intron | XP_011526682.1 | ||||
XM_011528382.2 | Intron | XP_011526684.1 | ||||
XM_011528383.1 | Intron | XP_011526685.1 | ||||
XM_011528384.2 | Intron | XP_011526686.1 | ||||
XM_011528385.2 | Intron | XP_011526687.1 | ||||
XM_017027401.1 | Intron | XP_016882890.1 |
Set Membership: |
HapMap |