Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606148 MIM: 600393 MIM: 608329 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FADS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
FADS1 - fatty acid desaturase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FEN1 - flap structure-specific endonuclease 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004111.5 | Intron | NP_004102.1 |
MIR611 - microRNA 611 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYRF - myelin regulatory factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM258 - transmembrane protein 258 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |