Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCTGCTCCTGTCACTGTCTTTT[C/T]ATAATGGCTTTTTCTGGCTCAGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604755 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372476 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC105372476 - uncharacterized LOC105372476 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF460 - zinc finger protein 460 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006635.3 | Intron | NP_006626.3 | ||||
XM_005258422.3 | Intron | XP_005258479.1 | ||||
XM_017026176.1 | Intron | XP_016881665.1 |
Set Membership: |
HapMap Validated |