Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCATTTTCTCTATAAAAGTCACA[C/G]TTTGTAACATATAGCCAACTATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108370 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASNS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ASNS - asparagine synthetase (glutamine-hydrolyzing) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178075.1 | Intron | NP_001171546.1 | ||||
NM_001178076.1 | Intron | NP_001171547.1 | ||||
NM_001178077.1 | Intron | NP_001171548.1 | ||||
NM_001673.4 | Intron | NP_001664.3 | ||||
NM_133436.3 | Intron | NP_597680.2 | ||||
NM_183356.3 | Intron | NP_899199.2 |
Set Membership: |
HapMap Validated |