Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAGTGAGACAGATCTGAGATTTA[A/C]ACACTTGAGCAGATGATAAAGCAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605073 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1E PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPM1E - protein phosphatase, Mg2+/Mn2+ dependent 1E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014906.4 | 4563 | Intron | NP_055721.3 | |||
XM_005257169.4 | 4563 | Intron | XP_005257226.1 | |||
XM_011524534.2 | 4563 | Intron | XP_011522836.1 | |||
XM_017024359.1 | 4563 | Intron | XP_016879848.1 |
Set Membership: |
HapMap |