Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGATCACATTTATTTTTAGAGG[C/T]GGAATTCAAAAAATAATAATGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605073 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1E PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPM1E - protein phosphatase, Mg2+/Mn2+ dependent 1E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014906.4 | Intron | NP_055721.3 | ||||
XM_005257169.4 | Intron | XP_005257226.1 | ||||
XM_011524534.2 | Intron | XP_011522836.1 | ||||
XM_017024359.1 | Intron | XP_016879848.1 |
Set Membership: |
HapMap |