Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCCTCTCCACTCCTCTGTGCTG[C/T]GGTGCTTAGCGGTTAATGGGCATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612420 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFAP1L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
VWA2 - von Willebrand factor A domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272046.1 | Intron | NP_001258975.1 | ||||
NM_001320804.1 | Intron | NP_001307733.1 | ||||
XM_011539754.2 | Intron | XP_011538056.1 | ||||
XM_011539757.2 | Intron | XP_011538059.1 | ||||
XM_017016177.1 | Intron | XP_016871666.1 | ||||
XM_017016178.1 | Intron | XP_016871667.1 | ||||
XM_017016179.1 | Intron | XP_016871668.1 | ||||
XM_017016180.1 | Intron | XP_016871669.1 | ||||
XM_017016181.1 | Intron | XP_016871670.1 |
Set Membership: |
HapMap Validated |