Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAGCCAGTGAGTCAGCCTTGGC[G/T]GAGCTTTTTCTGTGGCTTTTCCCAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607372 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED15 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
MED15 - mediator complex subunit 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003891.2 | Intron | NP_001003891.1 | ||||
NM_001293234.1 | Intron | NP_001280163.1 | ||||
NM_001293235.1 | Intron | NP_001280164.1 | ||||
NM_001293236.1 | Intron | NP_001280165.1 | ||||
NM_001293237.1 | Intron | NP_001280166.1 | ||||
NM_015889.4 | Intron | NP_056973.2 | ||||
XM_006724263.3 | Intron | XP_006724326.1 | ||||
XM_006724264.3 | Intron | XP_006724327.1 | ||||
XM_011530214.2 | Intron | XP_011528516.1 | ||||
XM_011530215.2 | Intron | XP_011528517.1 | ||||
XM_011530218.2 | Intron | XP_011528520.1 | ||||
XM_011530219.1 | Intron | XP_011528521.1 | ||||
XM_011530220.2 | Intron | XP_011528522.1 | ||||
XM_017028817.1 | Intron | XP_016884306.1 | ||||
XM_017028818.1 | Intron | XP_016884307.1 | ||||
XM_017028819.1 | Intron | XP_016884308.1 | ||||
XM_017028820.1 | Intron | XP_016884309.1 | ||||
XM_017028821.1 | Intron | XP_016884310.1 | ||||
XM_017028822.1 | Intron | XP_016884311.1 | ||||
XM_017028823.1 | Intron | XP_016884312.1 | ||||
XM_017028824.1 | Intron | XP_016884313.1 |