Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTTAAAAACAACACAAAGCATT[C/T]CAGTGACCCTATGAAGTCTTTTTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604430 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNG7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GNG7 - G protein subunit gamma 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052847.2 | 899 | UTR 3 | NP_443079.1 | |||
XM_017026606.1 | 899 | UTR 3 | XP_016882095.1 |
LOC101929097 - uncharacterized LOC101929097 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005259695.2 | 899 | Intron | XP_005259752.1 |
Set Membership: |
HapMap Validated |