Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGCCTTTTCCATGGCCCTGTTC[C/T]GAATGCTTTATGTGTGAGAACTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608134 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PALM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PALM - paralemmin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040134.1 | Intron | NP_001035224.1 | ||||
NM_002579.2 | Intron | NP_002570.2 | ||||
XM_005259565.4 | Intron | XP_005259622.1 | ||||
XM_005259566.4 | Intron | XP_005259623.1 | ||||
XM_017026850.1 | Intron | XP_016882339.1 |