Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATGTGTGTTTGTGTCTTTGTCAC[A/G]CATGCCTGGCATTGTGTTTTGCCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602656 MIM: 191092 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NTHL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NTHL1 - nth-like DNA glycosylase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSC2 - tuberous sclerosis 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000548.4 | Intron | NP_000539.2 | ||||
NM_001077183.2 | Intron | NP_001070651.1 | ||||
NM_001114382.2 | Intron | NP_001107854.1 | ||||
NM_001318827.1 | Intron | NP_001305756.1 | ||||
NM_001318829.1 | Intron | NP_001305758.1 | ||||
NM_001318831.1 | Intron | NP_001305760.1 | ||||
NM_001318832.1 | Intron | NP_001305761.1 | ||||
XM_005255529.4 | Intron | XP_005255586.2 | ||||
XM_005255531.4 | Intron | XP_005255588.2 | ||||
XM_011522636.2 | Intron | XP_011520938.1 | ||||
XM_011522637.2 | Intron | XP_011520939.1 | ||||
XM_011522638.2 | Intron | XP_011520940.2 | ||||
XM_011522639.2 | Intron | XP_011520941.1 | ||||
XM_011522640.2 | Intron | XP_011520942.1 | ||||
XM_017023615.1 | Intron | XP_016879104.1 | ||||
XM_017023616.1 | Intron | XP_016879105.1 | ||||
XM_017023617.1 | Intron | XP_016879106.1 | ||||
XM_017023618.1 | Intron | XP_016879107.1 |
Set Membership: |
HapMap |