Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147150 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MX1 - MX dynamin like GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144925.2 | 113 | UTR 5 | NP_001138397.1 | |||
NM_001178046.2 | 113 | Intron | NP_001171517.1 | |||
NM_001282920.1 | 113 | Intron | NP_001269849.1 | |||
NM_002462.4 | 113 | Intron | NP_002453.2 | |||
XM_005260978.4 | 113 | Intron | XP_005261035.1 | |||
XM_005260979.2 | 113 | Intron | XP_005261036.1 | |||
XM_005260980.2 | 113 | Intron | XP_005261037.1 | |||
XM_005260981.2 | 113 | Intron | XP_005261038.1 | |||
XM_005260982.2 | 113 | Intron | XP_005261039.1 | |||
XM_011529568.2 | 113 | UTR 5 | XP_011527870.1 | |||
XM_011529570.2 | 113 | Intron | XP_011527872.1 | |||
XM_017028349.1 | 113 | UTR 5 | XP_016883838.1 | |||
XM_017028350.1 | 113 | Intron | XP_016883839.1 | |||
XM_017028351.1 | 113 | Intron | XP_016883840.1 | |||
XM_017028352.1 | 113 | Intron | XP_016883841.1 |
Set Membership: |
HapMap |