Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACATTAAAACACACCCAAACTTGT[A/T]ATTTGGGTTTGCATCATTAGTTAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164740 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ETS2 - ETS proto-oncogene 2, transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256295.1 | Intron | NP_001243224.1 | ||||
NM_005239.5 | Intron | NP_005230.1 | ||||
XM_005260935.1 | Intron | XP_005260992.1 | ||||
XM_017028290.1 | Intron | XP_016883779.1 |
Set Membership: |
HapMap Validated |