Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAGCGTGTAGGAGTGATTTTCCT[C/T]GGAGCCTGTTTGTGGGTGGATGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607320 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM98C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAM98C - family with sequence similarity 98 member C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASGRP4 - RAS guanyl releasing protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146202.1 | 1970 | Missense Mutation | AAG,GAG | K,E 606 | NP_001139674.1 | |
NM_001146203.1 | 1970 | Missense Mutation | AAG,GAG | K,E 528 | NP_001139675.1 | |
NM_001146204.1 | 1970 | Missense Mutation | AAG,GAG | K,E 551 | NP_001139676.1 | |
NM_001146205.1 | 1970 | Missense Mutation | AAG,GAG | K,E 586 | NP_001139677.1 | |
NM_001146206.1 | 1970 | Missense Mutation | AAG,GAG | K,E 431 | NP_001139678.1 | |
NM_001146207.1 | 1970 | Missense Mutation | AAG,GAG | K,E 523 | NP_001139679.1 | |
NM_170604.2 | 1970 | Missense Mutation | AAG,GAG | K,E 620 | NP_733749.1 |
Set Membership: |
Validated |