Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116896 MIM: 605300 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CUX1 - cut like homeobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4285 - microRNA 4285 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH2B2 - SH2B adaptor protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020979.4 | 463 | Intron | NP_066189.3 | |||
XM_005276975.4 | 463 | UTR 5 | XP_005277032.1 | |||
XM_005276976.3 | 463 | UTR 5 | XP_005277033.1 | |||
XM_005276977.4 | 463 | Intron | XP_005277034.1 | |||
XM_005276979.3 | 463 | Intron | XP_005277036.1 | |||
XM_011515740.2 | 463 | Intron | XP_011514042.1 | |||
XM_017011680.1 | 463 | Intron | XP_016867169.1 | |||
XM_017011681.1 | 463 | Intron | XP_016867170.1 |
Set Membership: |
HapMap Validated |