Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGATGTGTTCCATTCTTTAAGCA[C/T]GTCCTTACTTTCTAGAACAATAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610138 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST6GALNAC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ST6GALNAC1 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289107.1 | 1737 | Intron | NP_001276036.1 | |||
NM_018414.4 | 1737 | Intron | NP_060884.1 | |||
XM_011524995.2 | 1737 | Silent Mutation | ACA,ACG | T,T 539 | XP_011523297.1 | |
XM_011524996.2 | 1737 | Intron | XP_011523298.1 | |||
XM_017024842.1 | 1737 | Silent Mutation | ACA,ACG | T,T 539 | XP_016880331.1 | |
XM_017024843.1 | 1737 | Intron | XP_016880332.1 | |||
XM_017024844.1 | 1737 | Intron | XP_016880333.1 |