Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600746 MIM: 606205 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDX1 - caudal type homeobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC6A7 - solute carrier family 6 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014228.4 | Intron | NP_055043.2 | ||||
XM_017009767.1 | Intron | XP_016865256.1 | ||||
XM_017009768.1 | Intron | XP_016865257.1 | ||||
XM_017009769.1 | Intron | XP_016865258.1 | ||||
XM_017009770.1 | Intron | XP_016865259.1 |
Set Membership: |
HapMap |