Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTTCCAGAGGCAAACACACTGG[C/T]CCATGAGTAGCAGAACGAGGGTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600856 MIM: 603240 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKN1C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDKN1C - cyclin dependent kinase inhibitor 1C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A18AS - solute carrier family 22 member 18 antisense | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302862.1 | 1406 | Intron | NP_001289791.1 | |||
NM_007105.3 | 1406 | Intron | NP_009036.2 | |||
XM_017017832.1 | 1406 | Intron | XP_016873321.1 | |||
XM_017017833.1 | 1406 | Intron | XP_016873322.1 | |||
XM_017017834.1 | 1406 | Intron | XP_016873323.1 | |||
XM_017017835.1 | 1406 | UTR 5 | XP_016873324.1 | |||
XM_017017836.1 | 1406 | Intron | XP_016873325.1 |