Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602986 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRG2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DRG2 - developmentally regulated GTP binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001388.4 | Intron | NP_001379.1 | ||||
XM_005256499.2 | Intron | XP_005256556.1 | ||||
XM_005256500.2 | Intron | XP_005256557.1 | ||||
XM_011523704.1 | Intron | XP_011522006.1 | ||||
XM_011523705.1 | Intron | XP_011522007.1 | ||||
XM_011523706.1 | Intron | XP_011522008.1 | ||||
XM_011523707.1 | Intron | XP_011522009.1 | ||||
XM_017024296.1 | Intron | XP_016879785.1 | ||||
XM_017024297.1 | Intron | XP_016879786.1 |