Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTGACATTTAAATCAAGACACG[C/T]TGCTTTTATTCCAAGTTGCACATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300566 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHFPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LHFPL1 - lipoma HMGIC fusion partner-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178175.3 | Intron | NP_835469.1 | ||||
XM_011530943.2 | Intron | XP_011529245.1 | ||||
XM_011530944.2 | Intron | XP_011529246.1 | ||||
XM_011530946.2 | Intron | XP_011529248.1 | ||||
XM_017029485.1 | Intron | XP_016884974.1 |