Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614133 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEPACAM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HEPACAM2 - HEPACAM family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039372.2 | Intron | NP_001034461.1 | ||||
NM_001288804.1 | Intron | NP_001275733.1 | ||||
NM_001288810.1 | Intron | NP_001275739.1 | ||||
NM_198151.2 | Intron | NP_937794.1 | ||||
XM_011516000.2 | Intron | XP_011514302.1 | ||||
XM_011516001.1 | Intron | XP_011514303.1 |