Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGAGGTGAGGAGTAGAAGTCAG[A/G]TTGGCAGGTGGGGGAGGCAACGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616582 MIM: 600390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LSR - lipolysis stimulated lipoprotein receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USF2 - upstream transcription factor 2, c-fos interacting | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321150.1 | Intron | NP_001308079.1 | ||||
NM_003367.3 | Intron | NP_003358.1 | ||||
NM_207291.2 | Intron | NP_997174.1 | ||||
XM_005259197.3 | Intron | XP_005259254.1 | ||||
XM_011527260.2 | Intron | XP_011525562.1 | ||||
XM_011527261.2 | Intron | XP_011525563.1 | ||||
XM_017027199.1 | Intron | XP_016882688.1 |