Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAACTGCATGCATATTATTCAGC[A/G]TCCTATATTCAAAGGTAACTACTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605094 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STEAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STEAP2 - STEAP2 metalloreductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040665.1 | 459 | Intron | NP_001035755.1 | |||
NM_001040666.1 | 459 | UTR 5 | NP_001035756.1 | |||
NM_001244944.1 | 459 | UTR 5 | NP_001231873.1 | |||
NM_001244945.1 | 459 | Intron | NP_001231874.1 | |||
NM_001244946.1 | 459 | Intron | NP_001231875.1 | |||
NM_152999.3 | 459 | Intron | NP_694544.2 | |||
XM_006715921.3 | 459 | UTR 5 | XP_006715984.1 | |||
XM_017011953.1 | 459 | UTR 5 | XP_016867442.1 | |||
XM_017011954.1 | 459 | Intron | XP_016867443.1 | |||
XM_017011955.1 | 459 | UTR 5 | XP_016867444.1 | |||
XM_017011956.1 | 459 | UTR 5 | XP_016867445.1 | |||
XM_017011957.1 | 459 | UTR 5 | XP_016867446.1 | |||
XM_017011958.1 | 459 | UTR 5 | XP_016867447.1 |
STEAP2-AS1 - STEAP2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |