Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP31 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
USP31 - ubiquitin specific peptidase 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020718.3 | 8112 | UTR 3 | NP_065769.3 | |||
XM_017023496.1 | 8112 | UTR 3 | XP_016878985.1 | |||
XM_017023497.1 | 8112 | UTR 3 | XP_016878986.1 | |||
XM_017023498.1 | 8112 | UTR 3 | XP_016878987.1 | |||
XM_017023499.1 | 8112 | UTR 3 | XP_016878988.1 | |||
XM_017023500.1 | 8112 | Intron | XP_016878989.1 |
Set Membership: |
HapMap |