Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300019 MIM: 312420 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCFC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HCFC1 - host cell factor C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005334.2 | 6861 | Silent Mutation | TGC,TGT | C,C 1953 | NP_005325.2 | |
XM_006724815.2 | 6861 | Silent Mutation | TGC,TGT | C,C 1998 | XP_006724878.1 | |
XM_006724816.2 | 6861 | Silent Mutation | TGC,TGT | C,C 1997 | XP_006724879.1 | |
XM_011531147.2 | 6861 | Silent Mutation | TGC,TGT | C,C 1957 | XP_011529449.1 | |
XM_011531148.2 | 6861 | Silent Mutation | TGC,TGT | C,C 1956 | XP_011529450.1 | |
XM_017029471.1 | 6861 | Silent Mutation | TGC,TGT | C,C 1931 | XP_016884960.1 | |
XM_017029472.1 | 6861 | Silent Mutation | TGC,TGT | C,C 1694 | XP_016884961.1 |
RENBP - renin binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |