Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171050 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ABCB1 - ATP binding cassette subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000927.4 | Intron | NP_000918.2 |
RUNDC3B - RUN domain containing 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134405.1 | Intron | NP_001127877.1 | ||||
NM_001134406.1 | Intron | NP_001127878.1 | ||||
NM_138290.2 | Intron | NP_612147.1 | ||||
XM_005250156.2 | Intron | XP_005250213.1 | ||||
XM_005250158.2 | Intron | XP_005250215.1 | ||||
XM_011515826.1 | Intron | XP_011514128.1 | ||||
XM_011515827.1 | Intron | XP_011514129.1 | ||||
XM_011515828.2 | Intron | XP_011514130.1 | ||||
XM_011515829.2 | Intron | XP_011514131.1 | ||||
XM_017011761.1 | Intron | XP_016867250.1 | ||||
XM_017011762.1 | Intron | XP_016867251.1 | ||||
XM_017011763.1 | Intron | XP_016867252.1 | ||||
XM_017011764.1 | Intron | XP_016867253.1 | ||||
XM_017011765.1 | Intron | XP_016867254.1 | ||||
XM_017011766.1 | Intron | XP_016867255.1 |
Set Membership: |
HapMap Validated |