Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCTTTCTTTCCAGGCGCAGCAA[A/G]CACACAGTGGTGGCCTATAAAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600574 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIFM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AIFM3 - apoptosis inducing factor, mitochondria associated 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LZTR1 - leucine zipper like transcription regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006767.3 | 313 | Silent Mutation | AAA,AAG | K,K 70 | NP_006758.2 |
Set Membership: |
HapMap |