Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300093 MIM: 300769 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRE PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
GABRE - gamma-aminobutyric acid type A receptor epsilon subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004961.3 | 1536 | UTR 3 | NP_004952.2 | |||
XM_011531140.2 | 1536 | UTR 3 | XP_011529442.1 | |||
XM_017029386.1 | 1536 | UTR 3 | XP_016884875.1 | |||
XM_017029387.1 | 1536 | UTR 3 | XP_016884876.1 | |||
XM_017029388.1 | 1536 | UTR 3 | XP_016884877.1 | |||
XM_017029389.1 | 1536 | UTR 3 | XP_016884878.1 |
MIR224 - microRNA 224 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR452 - microRNA 452 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |