Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTTGCAGGATAACCACAGTTGG[C/T]GACGCTTTCAGGCAGCTGGGAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605485 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS41 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
VPS41 - VPS41, HOPS complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014396.3 | 2886 | UTR 3 | NP_055211.2 | |||
NM_080631.3 | 2886 | UTR 3 | NP_542198.2 | |||
XM_017011988.1 | 2886 | UTR 3 | XP_016867477.1 |
Set Membership: |
HapMap Validated |