Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGAAGCAAATGTGGAGAGTTAA[A/G]ATTTAAATAGGATGCAACTTCCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF398 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF398 - zinc finger protein 398 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020781.3 | Intron | NP_065832.1 | ||||
NM_170686.2 | Intron | NP_733787.1 | ||||
XM_011516439.2 | Intron | XP_011514741.1 | ||||
XM_011516440.2 | Intron | XP_011514742.1 | ||||
XM_011516441.2 | Intron | XP_011514743.1 |
Set Membership: |
HapMap Validated |