Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTTCTCCTTCCTATCTCTATGG[A/T]TATGAGACATGGGAAAAATCGACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611546 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELOVL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ELOVL6 - ELOVL fatty acid elongase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130721.1 | Intron | NP_001124193.1 | ||||
NM_024090.2 | Intron | NP_076995.1 | ||||
XM_011532233.2 | Intron | XP_011530535.1 | ||||
XM_011532234.2 | Intron | XP_011530536.1 | ||||
XM_011532235.2 | Intron | XP_011530537.1 |
Set Membership: |
HapMap Validated |