Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTAAGGGCCTTTTCTTCTCTCC[A/G]GCCGAGTCTCATCTCTTCTTATTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300683 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEPT6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SEPT6 - septin 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015129.5 | 2426 | Intron | NP_055944.2 | |||
NM_145799.3 | 2426 | UTR 3 | NP_665798.1 | |||
NM_145800.3 | 2426 | Intron | NP_665799.1 | |||
NM_145802.3 | 2426 | Intron | NP_665801.1 | |||
XM_005262400.3 | 2426 | UTR 3 | XP_005262457.1 | |||
XM_006724748.2 | 2426 | UTR 3 | XP_006724811.1 | |||
XM_006724750.2 | 2426 | Intron | XP_006724813.1 | |||
XM_011531317.2 | 2426 | Intron | XP_011529619.1 | |||
XM_011531318.2 | 2426 | Intron | XP_011529620.1 |
Set Membership: |
HapMap |