Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTCCTCTTCCCCCATCTTTCCAC[A/G]CTGAAGAGAGCATCTCAAGTCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600439 MIM: 614084 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SSBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SSBP1 - single stranded DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256510.1 | Intron | NP_001243439.1 | ||||
NM_001256511.1 | Intron | NP_001243440.1 | ||||
NM_001256512.1 | Intron | NP_001243441.1 | ||||
NM_001256513.1 | Intron | NP_001243442.1 | ||||
NM_003143.2 | Intron | NP_003134.1 | ||||
XM_005250048.4 | Intron | XP_005250105.1 | ||||
XM_005250049.4 | Intron | XP_005250106.1 | ||||
XM_005250050.4 | Intron | XP_005250107.1 | ||||
XM_005250051.4 | Intron | XP_005250108.1 |
WEE2 - WEE1 homolog 2 (S. pombe) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WEE2-AS1 - WEE2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |