Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACAGGCCTGGGCACTCACCCAC[A/G]TAGCTGAGGATGACGGGCAGGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608010 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPC1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NPC1L1 - NPC1 like intracellular cholesterol transporter 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101648.1 | 3447 | Silent Mutation | TAC,TAT | Y,Y 1264 | NP_001095118.1 | |
NM_001300967.1 | 3447 | Intron | NP_001287896.1 | |||
NM_013389.2 | 3447 | Silent Mutation | TAC,TAT | Y,Y 1291 | NP_037521.2 | |
XM_011515326.2 | 3447 | Silent Mutation | TAC,TAT | Y,Y 1199 | XP_011513628.1 | |
XM_011515327.2 | 3447 | Intron | XP_011513629.1 | |||
XM_011515328.2 | 3447 | Silent Mutation | TAC,TAT | Y,Y 717 | XP_011513630.1 |