Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600136 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MAP3K9 - mitogen-activated protein kinase kinase kinase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284230.1 | 9015 | UTR 3 | NP_001271159.1 | |||
NM_001284231.1 | 9015 | UTR 3 | NP_001271160.1 | |||
NM_001284232.1 | 9015 | UTR 3 | NP_001271161.1 | |||
NM_033141.3 | 9015 | UTR 3 | NP_149132.2 | |||
XM_005267683.4 | 9015 | UTR 3 | XP_005267740.1 | |||
XM_011536788.2 | 9015 | UTR 3 | XP_011535090.1 | |||
XM_011536792.1 | 9015 | UTR 3 | XP_011535094.1 | |||
XM_011536794.2 | 9015 | UTR 3 | XP_011535096.1 |
Set Membership: |
HapMap Validated |