Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609373 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM3B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KDM3B - lysine demethylase 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016604.3 | 179 | Silent Mutation | ATC,ATT | I,I 88 | NP_057688.2 | |
XM_005272018.4 | 179 | Silent Mutation | ATC,ATT | I,I 88 | XP_005272075.1 | |
XM_011543488.2 | 179 | Silent Mutation | ATC,ATT | I,I 44 | XP_011541790.1 | |
XM_011543489.2 | 179 | Silent Mutation | ATC,ATT | I,I 40 | XP_011541791.1 | |
XM_017009584.1 | 179 | Intron | XP_016865073.1 |
Set Membership: |
HapMap |